Assay Details
| Gene Symbol: GSPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098071.1 | NP_001091540.2 | 10 - 11 | 1581 | 87 | |
| GenBank mRNA | BC134651.1 | - | 10 - 11 | 1355 | 87 | |
| BC140515.1 | - | 10 - 11 | 1581 | 87 | ||
More Information
Set Membership: |
|

GSPT1
Genomic Map