Assay Details
| Gene Symbol: CNOT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101167.2 | NP_001094637.1 | 2 - 3 | 318 | 75 | |
| GenBank mRNA | BC151334.1 | - | 2 - 3 | 319 | 75 | |
More Information
Set Membership: |
|

CNOT9
Genomic Map