Assay Details
Gene Symbol: ETF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076254.1 | NP_001069722.1 | 2 - 3 | 357 | 116 | |
GenBank mRNA | AY911346.1 | - | 3 - 4 | 390 | 116 | |
BC119947.1 | - | 2 - 3 | 357 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
