Assay Details
| Gene Symbol: ARPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034271.2 | NP_001029443.1 | 6 - 7 | 510 | 114 | |
| GenBank mRNA | BC102590.1 | - | 6 - 7 | 511 | 114 | |
More Information
Set Membership: |
|

ARPC3
Genomic Map