Assay Details
| Gene Symbol: NECTIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024494.1 | NP_001019665.1 | 6 - 7 | 1412 | 75 | |
| GenBank mRNA | BC148055.1 | - | 6 - 7 | 1409 | 75 | |
| BT020770.1 | - | 6 - 7 | 1412 | 75 | ||
More Information
Set Membership: |
|

NECTIN4
Genomic Map