Assay Details
| Gene Symbol: CHN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075349.1 | NP_001068817.1 | 2 - 3 | 728 | 99 | |
| NM_001113247.1 | NP_001106718.1 | 9 - 10 | 1058 | 99 | ||
| GenBank mRNA | BC118265.1 | - | 2 - 3 | 728 | 99 | |
| BC126673.1 | - | 9 - 10 | 1052 | 99 | ||
| BC153235.1 | - | 9 - 10 | 1058 | 99 | ||
More Information
Set Membership: |
|

CHN1
Genomic Map