Assay Details
| Gene Symbol: CAV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174004.3 | NP_776429.1 | 2 - 3 | 256 | 67 | |
| GenBank mRNA | AY823915.1 | - | 2 - 3 | 256 | 67 | |
| BC103050.1 | - | 2 - 3 | 219 | 67 | ||
| U86639.1 | - | 2 - 3 | 194 | 67 | ||
More Information
Set Membership: |
|

CAV1
Genomic Map