Assay Details
| Gene Symbol: FGF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079789.1 | NP_001073257.1 | 5 - 6 | 594 | 103 | |
| GenBank mRNA | BC118169.1 | - | 5 - 6 | 594 | 103 | |
More Information
Set Membership: |
|

FGF12
Genomic Map