Assay Details
| Gene Symbol: AEBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083485.3 | NP_001076954.2 | 8 - 9 | 1489 | 100 | |
| GenBank mRNA | BC123767.1 | - | 8 - 9 | 1196 | 100 | |
More Information
Set Membership: |
|

AEBP2
Genomic Map