Assay Details
| Gene Symbol: CXXC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038176.2 | NP_001033265.2 | 2 - 3 | 1108 | 94 | |
| GenBank mRNA | EE223275.1 | - | 1 - 2 | 624 | 94 | |
More Information
Set Membership: |
|

CXXC5
Genomic Map