Assay Details
| Gene Symbol: CHTOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035481.2 | NP_001030558.1 | 3 - 4 | 464 | 71 | |
| GenBank mRNA | BC103347.1 | - | 3 - 4 | 465 | 71 | |
More Information
Set Membership: |
|

CHTOP
Genomic Map