Assay Details
| Gene Symbol: MYL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175780.1 | NP_786974.1 | 6 - 7 | 502 | 79 | |
| GenBank mRNA | BC142278.1 | - | 6 - 7 | 489 | 79 | |
| X54978.1 | - | 502 | 79 | |||
More Information
Set Membership: |
|

MYL6
Genomic Map