Assay Details
| Gene Symbol: SENP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191374.2 | NP_001178303.1 | 4 - 5 | 1216 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SENP3| Gene Symbol: SENP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191374.2 | NP_001178303.1 | 4 - 5 | 1216 | 81 | |
Set Membership: |
|