Assay Details
| Gene Symbol: SLC35D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082459.2 | NP_001075928.1 | 5 - 6 | 569 | 106 | |
| GenBank mRNA | BC133580.1 | - | 5 - 6 | 570 | 106 | |
More Information
Set Membership: |
|

SLC35D1
Genomic Map