Assay Details
Gene Symbol: GORASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038535.2 | NP_001033624.2 | 5 - 6 | 728 | 113 | |
GenBank mRNA | BC109691.1 | - | 5 - 6 | 581 | 113 | |
EE350057.1 | - | 5 - 6 | 577 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
