Assay Details
| Gene Symbol: UPRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076245.2 | NP_001069713.1 | 2 - 3 | 482 | 80 | |
| GenBank mRNA | BC109679.1 | - | 2 - 3 | 485 | 80 | |
More Information
Set Membership: |
|

UPRT
Genomic Map