Assay Details
| Gene Symbol: UBE2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046496.1 | NP_001039961.1 | 3 - 4 | 603 | 102 | |
| GenBank mRNA | AY291287.1 | - | 2 - 3 | 59 | 102 | |
| BC114654.1 | - | 3 - 4 | 603 | 102 | ||
More Information
Set Membership: |
|

UBE2D2
Genomic Map