Assay Details
| Gene Symbol: BSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001145.1 | NP_001001145.1 | 3 - 4 | 276 | 133 | |
| GenBank mRNA | M22244.1 | - | 3 - 4 | 276 | 133 | |
More Information
Set Membership: |
|

BSP1
Genomic Map