Assay Details
| Gene Symbol: MOGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001153.2 | NP_001001153.1 | 3 - 4 | 479 | 65 | |
| GenBank mRNA | AJ519785.2 | - | 3 - 4 | 479 | 65 | |
More Information
Set Membership: |
|

MOGAT1
Genomic Map