Assay Details
| Gene Symbol: P4HA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001598.2 | NP_001001598.1 | 8 - 9 | 1219 | 62 | |
| GenBank mRNA | AB126035.1 | - | 8 - 9 | 1220 | 62 | |
More Information
Set Membership: |
|

P4HA3
Genomic Map