Assay Details
| Gene Symbol: ZIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002888.1 | NP_001002888.1 | 1 - 2 | 463 | 97 | |
| GenBank mRNA | AY427789.1 | - | 1 - 2 | 463 | 97 | |
More Information
Set Membership: |
|

ZIM2
Genomic Map