Assay Details
| Gene Symbol: TUBB2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003900.1 | NP_001003900.1 | 1 | 99 | 57 | |
| GenBank mRNA | AY675081.1 | - | 1 | 99 | 57 | |
| BC118413.1 | - | 1 | 30 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118413.1, NM_001003900.1 transcripts.
Set Membership: |
|

TUBB2B
Genomic Map