Assay Details
Gene Symbol: PNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007818.1 | NP_001007819.1 | 1 - 2 | 103 | 75 | |
GenBank mRNA | AF529136.1 | - | 1 - 2 | 103 | 75 | |
BC103291.1 | - | 1 - 2 | 100 | 75 | ||
BT021545.1 | - | 1 - 2 | 99 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC103291.1, NM_001007818.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
