Assay Details
| Gene Symbol: KLK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008416.1 | NP_001008416.1 | 1 - 2 | 87 | 92 | |
| GenBank mRNA | AY290705.1 | - | 1 - 2 | 87 | 92 | |
| BC151558.1 | - | 1 - 2 | 82 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151558.1, NM_001008416.1 transcripts.
Set Membership: |
|

KLK1
Genomic Map