Assay Details
Gene Symbol: KRT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008663.1 | NP_001008663.1 | 4 - 5 | 933 | 105 | |
GenBank mRNA | BC140616.1 | - | 4 - 5 | 1034 | 105 | |
BC149167.1 | - | 3127 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
