Assay Details
| Gene Symbol: KRT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008663.1 | NP_001008663.1 | 6 - 7 | 1227 | 72 | |
| GenBank mRNA | BC140616.1 | - | 6 - 7 | 1328 | 72 | |
| BC149167.1 | - | 3421 | 72 | |||
More Information
Set Membership: |
|

KRT5
Genomic Map