Assay Details
| Gene Symbol: BHMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011679.1 | NP_001011679.1 | 6 - 7 | 863 | 66 | |
| GenBank mRNA | AY854632.1 | - | 6 - 7 | 863 | 66 | |
| BC112599.1 | - | 6 - 7 | 875 | 66 | ||
More Information
Set Membership: |
|

BHMT
Genomic Map