Assay Details
| Gene Symbol: STAT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012673.1 | NP_001012691.1 | 10 - 11 | 1327 | 54 | |
| GenBank mRNA | Z72482.1 | - | 10 - 11 | 1327 | 54 | |
More Information
Set Membership: |
|

STAT5A
Genomic Map