Assay Details
| Gene Symbol: PAK1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014852.1 | NP_001014852.1 | 5 - 6 | 527 | 73 | |
| GenBank mRNA | BC102508.1 | - | 5 - 6 | 526 | 73 | |
| BT020668.1 | - | 5 - 6 | 527 | 73 | ||
| BT020670.1 | - | 5 - 6 | 527 | 73 | ||
More Information
Set Membership: |
|

PAK1IP1
Genomic Map