Assay Details
Gene Symbol: MRPL49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014871.1 | NP_001014871.1 | 2 - 3 | 237 | 99 | |
GenBank mRNA | BC120239.1 | - | 2 - 3 | 230 | 99 | |
BT020698.1 | - | 2 - 3 | 237 | 99 | ||
BT026163.1 | - | 2 - 3 | 200 | 99 | ||
BT026181.1 | - | 2 - 3 | 211 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
