Assay Details
| Gene Symbol: RBM14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014901.1 | NP_001014901.1 | 1 - 2 | 418 | 88 | |
| GenBank mRNA | BC113267.1 | - | 1 - 2 | 412 | 88 | |
| BT020733.1 | - | 1 - 2 | 418 | 88 | ||
More Information
Set Membership: |
|

RBM14
Genomic Map