Assay Details
| Gene Symbol: SLC39A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015521.1 | NP_001015521.1 | 2 - 3 | 376 | 66 | |
| GenBank mRNA | BC122570.1 | - | 1 - 2 | 177 | 66 | |
| BT020718.1 | - | 2 - 3 | 376 | 66 | ||
| BT021060.1 | - | 2 - 3 | 135 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC122570.1, NM_001015521.1 transcripts.
Set Membership: |
|

SLC39A3
Genomic Map