Assay Details
| Gene Symbol: RIOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015675.1 | NP_001015675.1 | 4 - 5 | 818 | 73 | |
| GenBank mRNA | BC109961.1 | - | 4 - 5 | 744 | 73 | |
| BT020745.1 | - | 4 - 5 | 818 | 73 | ||
More Information
Set Membership: |
|

RIOX2
Genomic Map