Assay Details
| Gene Symbol: RIOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015675.1 | NP_001015675.1 | 7 - 8 | 1196 | 82 | |
| GenBank mRNA | BT020745.1 | - | 7 - 8 | 1196 | 82 | |
More Information
Set Membership: |
|

RIOX2
Genomic Map