Assay Details
| Gene Symbol: LRRC51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024545.1 | NP_001019716.1 | 3 - 4 | 297 | 86 | |
| NM_001282038.1 | NP_001268967.1 | 3 - 4 | 258 | 86 | ||
| NM_001282039.1 | NP_001268968.1 | 2 - 3 | 197 | 86 | ||
| GenBank mRNA | BC109501.1 | - | 2 - 3 | 198 | 86 | |
| BT020631.1 | - | 3 - 4 | 297 | 86 | ||
| EE348175.1 | - | 3 - 4 | 258 | 86 | ||
More Information
Set Membership: |
|

LRRC51
Genomic Map