Assay Details
| Gene Symbol: ATP6V0E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034050.2 | NP_001029222.1 | 3 - 4 | 338 | 78 | |
| GenBank mRNA | BC103204.1 | - | 3 - 4 | 339 | 78 | |
| Y15285.1 | - | 3 - 4 | 229 | 78 | ||
More Information
Set Membership: |
|

ATP6V0E1
Genomic Map