Assay Details
Gene Symbol: CD1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034394.2 | NP_001029566.1 | 1 - 2 | 120 | 114 | |
GenBank mRNA | BC102922.1 | - | 1 - 2 | 121 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
