Assay Details
| Gene Symbol: CD1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034394.2 | NP_001029566.1 | 2 - 3 | 413 | 99 | |
| GenBank mRNA | BC102922.1 | - | 2 - 3 | 414 | 99 | |
More Information
Set Membership: |
|

CD1E
Genomic Map