Assay Details
| Gene Symbol: ARFIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034477.2 | NP_001029649.1 | 3 - 4 | 350 | 87 | |
| GenBank mRNA | BC102052.1 | - | 3 - 4 | 351 | 87 | |
| BT025433.1 | - | 2 - 3 | 251 | 87 | ||
More Information
Set Membership: |
|

ARFIP2
Genomic Map