Assay Details
Gene Symbol: NFYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034598.1 | NP_001029770.1 | 7 - 8 | 889 | 56 | |
GenBank mRNA | BC105387.1 | - | 7 - 8 | 877 | 56 | |
BT020742.1 | - | 7 - 8 | 889 | 56 | ||
BT020799.1 | - | 7 - 8 | 885 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
