Assay Details
| Gene Symbol: MRPL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034608.2 | NP_001029780.1 | 2 - 3 | 345 | 60 | |
| GenBank mRNA | BC103149.1 | - | 2 - 3 | 346 | 60 | |
More Information
Set Membership: |
|

MRPL18
Genomic Map