Assay Details
| Gene Symbol: MOB3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034755.1 | NP_001029927.1 | 4 - 5 | 2473 | 107 | |
| GenBank mRNA | BT020665.1 | - | 4 - 5 | 2473 | 107 | |
More Information
Set Membership: |
|

MOB3C
Genomic Map