Assay Details
| Gene Symbol: DHRS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035088.2 | NP_001030260.1 | 2 - 3 | 428 | 61 | |
| GenBank mRNA | BC103081.1 | - | 2 - 3 | 430 | 61 | |
More Information
Set Membership: |
|

DHRS11
Genomic Map