Assay Details
| Gene Symbol: GPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035354.1 | NP_001030431.1 | 2 - 3 | 259 | 58 | |
| GenBank mRNA | BC105513.1 | - | 2 - 3 | 206 | 58 | |
| BT020681.1 | - | 2 - 3 | 259 | 58 | ||
More Information
Set Membership: |
|

GPD1
Genomic Map