Assay Details
Gene Symbol: RABEPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035399.1 | NP_001030476.1 | 4 - 5 | 544 | 69 | |
GenBank mRNA | BC111169.1 | - | 5 - 6 | 570 | 69 | |
BT020719.1 | - | 4 - 5 | 544 | 69 | ||
BT021185.1 | - | 4 - 5 | 546 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
