Assay Details
| Gene Symbol: SEC61A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040504.1 | NP_001035594.1 | 7 - 8 | 644 | 64 | |
| GenBank mRNA | BC151365.1 | - | 7 - 8 | 683 | 64 | |
| BT020701.1 | - | 7 - 8 | 644 | 64 | ||
More Information
Set Membership: |
|

SEC61A1
Genomic Map