Assay Details
| Gene Symbol: IFI6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075588.1 | NP_001069056.1 | 1 - 2 | 56 | 67 | |
| GenBank mRNA | BC111668.1 | - | 1 - 2 | 79 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111668.1, NM_001075588.1 transcripts.
Set Membership: |
|

IFI6
Genomic Map