Assay Details
Gene Symbol: IFI6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075588.1 | NP_001069056.1 | 2 - 3 | 145 | 80 | |
GenBank mRNA | BC111668.1 | - | 2 - 3 | 168 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
