Assay Details
Gene Symbol: IFI6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075588.1 | NP_001069056.1 | 4 - 5 | 380 | 128 | |
GenBank mRNA | BC111668.1 | - | 4 - 5 | 403 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
