Assay Details
Gene Symbol: TAFA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077000.2 | NP_001070468.1 | 2 - 3 | 321 | 92 | |
GenBank mRNA | BC103140.1 | - | 2 - 3 | 322 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
